

硼酸三甲酯 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:121-43-7
:
Formula:C₃H₉BO₃
MDL Number:MFCD00008346
:103.91 g/mol
Merck Index:12,09842
Boiling Pt:68…69 °C (760 mmHg)
Melting Pt:–34 °C
Density:0,915 g/cm³
Flash Pt:–8 °C
Storage Temperature:Ambient
EINECS:204-468-9
UN:2416
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3562 to 1.3582 (20°C, 589 nm)
- Titration with NaOH:≥98.5 %