Order Entry
China
ContactUsLinkComponent
 
 
Triethyl borate 97%
  ACRO139582500
 :  
undefined
 
Triethyl borate 97%
  ACRO139582500
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

 

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3705 to 1.3775 (20°C, 589 nm)
  • GC:
    ≥96.0 %

 

 

 :
 :145.99 g/mol
 :117…118 °C (760 mmHg)
 :–84.5 °C
 :0.858 g/cm³
 :11 °C
 :Ambient
 :205-760-9
 :1176
 :3,II
 :GHS02
 :150-46-9
 :C₆H₁₅BO₃
 :MFCD00012846