Magnesium oxide 96.0-100.5% (excluding ignited substances), powder FCC, USP, J.T.Baker®
: Avantor
:1309-48-4
:
:MgO
:MFCD00011109
:40.3 g/mol
:3600 °C (1013 hPa)
:2800 °C
:3.58 g/cm³ (25 °C)
:Management of Change (MOC) category = R
:215-171-9
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Assay (MgO) (ignited basis):96.0 - 100.5 %
- USP - Loss on Ignition:≤ 10.0 %
- USP - Free Alkali and Soluble Salts:≤ 2.0 %
- USP - Acid-Insoluble Substances:≤ 0.1 %
- USP - Bulk Density, For Information Only:≤ 0.05 %
- USP - Calcium (Ca):≤ 1.1 %
- USP - Iron (Fe):≤ 0.05 %
- FCC - Identification:Passes Test
- FCC - Assay (MgO) (ignited basis):96.0 - 100.5 %
- FCC - Loss on Ignition:≤ 10.0 %
- FCC - Free Alkali and Soluble Salts:≤ 2.0 %
- FCC - Acid-Insoluble Substances:≤ 0.1 %
- FCC - Arsenic (As):≤ 3 mg/kg
- FCC - Calcium Oxide:≤ 1.5 %
- FCC - Lead (Pb):≤ 4 mg/kg