

Sodium hydroxide solution 10 N, Biotech Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:1.32 g/cm³
Storage Temperature:Ambient
EINECS:231-820-9
UN:1824
ADR:8,II
:GHS05
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- GMP Manufactured Product:9.9 - 10.1
- Store above 16°C (60°F) to prevent freezing:≤0.6%
- CAUTION: For Manufacturing, processing or repackaging:≤5 ppm
- Appearance (Clear, colorless liquid):Passes Test
- Normality:9.9 - 10.1
- Sodium Carbonate (Na₂CO₃):≤0.6 %
- Chloride (Cl):≤20 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤2 ppm
- Endotoxin Concentration (EU/mL):≤2.0
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Cell Culture Solutions.pdf
Product Literature
spec_BAKR0312-02 spec_BAKR0312-07