Order Entry
China
ContactUsLinkComponent
Sodium hydrogen carbonate, Sigma-Aldrich®
Sodium hydrogen carbonate, Sigma-Aldrich®
Catalog # SIALS6297-250G
Supplier:  SIGMAALDRICH
CAS Number:  
undefined
Sodium hydrogen carbonate, Sigma-Aldrich®
Catalog # SIALS6297-250G
Supplier:  SIGMAALDRICH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.159 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:205-633-8
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528