
Sodium tetraphenylborate 99.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:143-66-8
:
Formula:(C₆H₅)₄BNa
MDL Number:MFCD00011494
:342.22 g/mol
Merck Index:13,08765
Melting Pt:>300 °C
Density:1,15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-605-5
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to light beige Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C)
- Titration with HClO4:99.4 to 100.6 %