Order Entry
China
ContactUsLinkComponent
 
Irritant
Phenol red, pure indicator
Catalog # ACRO151430250
CAS Number:  
undefined
Irritant
Phenol red, pure indicator
Catalog # ACRO151430250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Red Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in 95% ethanol) Clear soluble
  • UV Lambda max:
    555 to 561 nm (in buffer)
  • Specific extinction E (1%/1cm):
    ≥1700 (at lambda max., in buffer)
  • Visual transition interval:
    from pH 6.8 (yellow) to pH 8.2 (red)

Specification Test Results

About this item

 :
Merck Index:13,07329
Melting Pt:>300 °C
 :354.38 g/mol
EINECS:205-609-7
 :GHS07
Storage Temperature:Ambient
CAS Number:143-74-8
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552