Order Entry
China
ContactUsLinkComponent
 
Bromothymol blue, pure indicator
 
undefined
Bromothymol blue, pure indicator
 

 

  • Appearance:
    Light pink to brown to purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies
  • Solubility:
    (0.1 % in ethanol) Clear
  • Visual transition interval:
    from pH 6.0 (yellow) to pH 7.6 (blue)