
Ethyl-4-fluorobenzoate 99%
:451-46-7
:
:C₉H₉FO₂
:MFCD00000351
:168.17 g/mol
:13,04198
:210 °C (760 mmHg)
:25…27 °C
:1.14 g/cm³
:81 °C
:207-194-8
:GHS07
:Ambient
- Appearance:Colorless to light yellow Low melting solid
- Infrared spectrum:Conforms
- Refractive index:1.4850 to 1.4870 (20°C, 589 nm)
- GC:≥98.5 %