Order Entry
China
Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ MabSelect™ PrismA

Affinity chromatography columns, HiTrap™ MabSelect™ PrismA

HiTrap™ MabSelect™ PrismA, PP, 60 µm, 7×25 mm, 1 ml, <4 ml/min
Catalog # GEHE17-5498-52
Supplier:   GE HEALTHCARE
Affinity chromatography columns, HiTrap™ MabSelect™ PrismA
Catalog # GEHE17-5498-52
Supplier:  GE HEALTHCARE
Affinity chromatography columns, HiTrap™ MabSelect™ PrismA
Affinity chromatography columns, HiTrap™ MabSelect™ PrismA

HiTrap™ MabSelect™ PrismA, PP, 60 µm, 7×25 mm, 1 ml, <4 ml/min

Price
Inquire For Price
Log in for availability
1 * 5 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These columns are prepacked with MabSelect™ PrismA protein A chromatographyn resin that has been improved with an optimised high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs. These columns are suitable for purifying antibodies with affinity chromatography in research and process development.

  • Enhanced binding capacity compared with other Protein A resins
  • Excellent alkaline stability enables efficient cleaning and sanitisation using 0,5 to 1,0 M NaOH
  • Convenient HiTrap™ format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system

MabSelect™ PrismA builds on the proven track record of MabSelect™ and MabSelect™ SuRe™ resins in commercial mAb production.

The columns are made of biocompatible PP that does not interact with biomolecules.

Caution: Note that HiTrap™ columns cannot be opened or repacked.

Specifications
  • Flow rate:
    <4 ml/min
  • Particle Size:
    60 µm
  • Capacity:
    1 ml
  • Description:
    HiTrap™ MabSelect™ PrismA
  • Seq:
    02
  • Column hardware:
    PP
  • Bed dimensions:
    7×25 mm
  • Cat. No.:
    GEHE17-5498-52
  • Max. pressure:
    5 bar
  • Binding capacity:
    ~ 40 mg polyclonal IgG/ml resin, 2 min residence time
    ~ 80 mg polyclonal IgG/ml resin, 6 min residence time
  • Storage:
    20% ethanol, 2 to 8 °C
  • Temperature limit:
    40 °C
  • pH range:
    3 – 12

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.