
Methyl-4-bromobenzoate 99%
:619-42-1
:
:C₈H₇BrO₂
:MFCD00013531
:215.05 g/mol
:13,01392
:246 °C (1013 hPa)
:78…81 °C
:1,689 g/cm³ (20 °C)
:210-596-6
:GHS07
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:78 to 81 °C
- GC:≥98.5 %