Order Entry
China
ContactUsLinkComponent
Isoamyl alcohol ≥98% for milk analysis according to the NF V 04-210 standard
Isoamyl alcohol ≥98% for milk analysis according to the NF V 04-210 standard
Catalog # VWRC20799.298
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Isoamyl alcohol ≥98% for milk analysis according to the NF V 04-210 standard
Catalog # VWRC20799.298
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Density (20/4):
    0.808 - 0.818
  • Assay (calculated as mixture of isomers):
    Min. 98 %
  • Fats:
    Passes test
  • Furfuraldehyde:
    Passes test
  • Organic impurities:
    Passes test
  • Distillation range:
    128 - 132 °C
  • Evaporation residue:
    Max. 50 ppm
  • 2-Methyl-1-butanol:
    8 - 10 %
  • 3-Methyl-1-butanol:
    90 - 92 %
  • Water:
    Max. 0.5 %

Specification Test Results

About this item

 :
 :88.15 g/mol
Merck Index:13,05214
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–117 °C
Density:0.812 g/cm³ (20 °C)
Flash Pt:42 °C
Storage Temperature:Ambient
EINECS:204-633-5
UN:1105
ADR:3,III
 :GHS02
 :GHS07
CAS Number:123-51-3
Formula:(CH₃)₂CHCH₂CH₂OH
MDL Number:MFCD00002934