

Isoamyl alcohol, dehydrated (max. 0.05% H₂O) ≥98%, GPR RECTAPUR® for synthesis
Catalog # VWRC20796.298
Supplier: VWR Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay (calculated as mixture of isomers):Min. 98 %
- Density (20/4):0.808 - 0.812
- Water:Max. 0.05 %
Specification Test Results
About this item
:
:88.15 g/mol
Merck Index:13,05214
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–117 °C
Density:0.812 g/cm³ (20 °C)
Flash Pt:42 °C
Storage Temperature:Ambient
EINECS:204-633-5
UN:1105
ADR:3,III
:GHS02
:GHS07
CAS Number:123-51-3
Formula:(CH₃)₂CHCH₂CH₂OH
MDL Number:MFCD00002934