

Isoamyl alcohol ACS
Catalog # ACRO412735000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.5 %
- GC:≥98.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.003 %
- Carbonyl (as HCHO):≤0.1 %
- Acids and esters:≤0.2 % (as Amyl acetate)
- Titrable acid:≤0.002 meq/g
Specification Test Results
About this item
:
:88.15 g/mol
Merck Index:13,05214
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.8 g/cm³
Flash Pt:45 °C
Storage Temperature:Ambient
EINECS:204-633-5
UN:1105
ADR:3,III
:GHS07
:GHS02
CAS Number:123-51-3
Formula:(CH₃)₂CHCH₂CH₂OH
MDL Number:MFCD00002934