
1-Hexanesulphonic acid sodium salt monohydrate 98% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207300-91-2
:
Formula:C₆H₁₃NaO₃S·1H₂O
MDL Number:MFCD00149549
Melting Pt:>300 °C
:206.24 g/mol
EINECS:220-601-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:7 to 9 %
- Solubility:(10 % in water) Clear colorless
- UV:(0.25 M in water) 1 cm cell vs HPLC-grade water
- Titration after Ion exchange:≥97.5 % (On dry substance)