Order Entry
China
ContactUsLinkComponent
 
Irritant
Flammable
Lithium dodecyl sulphate 99+%
Catalog # ACRO413300250
CAS Number:  
undefined
Irritant
Flammable
Lithium dodecyl sulphate 99+%
Catalog # ACRO413300250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White Glistening crystalline powder or flakes
  • Infrared spectrum:
    Conforms
  • UV:
    (5 % in water) 1 cm cell
  • Titration after Ion exchange:
    ≥99.0 %

Specification Test Results

About this item

 :
 :272.34 g/mol
Storage Temperature:Ambient
EINECS:218-058-2
UN:1325
ADR:4.1,III
 :GHS07
 :GHS02
CAS Number:2044-56-6
Formula:C₁₂H₂₅LiO₄S
MDL Number:MFCD00007467