Order Entry
China
ContactUsLinkComponent
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
  MERP1.37028.1000
 :  
undefined
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
  MERP1.37028.1000
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024
 :
 :92.09 g/mol
 :13,04497
 :290 °C (1013 hPa)
 :18.6 °C
 :1.26 g/cm³ (20 °C)
 :177 °C
 :Ambient
 :200-289-5
 :56-81-5
 :HOCH₂CH(OH)CH₂OH
 :MFCD00004722