Order Entry
China
ContactUsLinkComponent
1,6-Hexanediol 97%
1,6-Hexanediol 97%
Certificates
undefined
1,6-Hexanediol 97%

Specification Test Results

  • Appearance:
    Colorless to white to light yellow Solidified melt or liquid
  • Infrared spectrum:
    Conforms
  • Melting point:
    40 to 44 °C
  • Water:
    ≤0.3 % (K.F.)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information