

Bis(2-methoxyethyl)ether 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-96-6
:
Formula:C₆H₁₄O₃
MDL Number:MFCD00008503
Merck Index:13,03187
Boiling Pt:162 °C (760 mmHg)
Melting Pt:–64 °C
Density:0.937 g/cm³
Flash Pt:51 °C
UN:3271
ADR:3,III
:134.18 g/mol
EINECS:203-924-4
:GHS08
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4070 to 1.4090 (20°C, 589 nm)
- Water:≤0.025 %
- GC:≥98.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- Acidity (CH3COOH):≤0.015 %
- Peroxides:≤0.005 %