


1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Catalog # ACRO610351000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
- Stabilizer:Approx. 100 ppm BHT
- Residue after ignition:≤0.001 %
Specification Test Results
About this item
:
:90.12 g/mol
Merck Index:13,03251
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–69 °C
Density:0.867 g/cm³
Flash Pt:–6 °C
EINECS:203-794-9
UN:2252
ADR:3,II
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
CAS Number:110-71-4
Formula:C₄H₁₀O₂
MDL Number:MFCD00008502