Order Entry
China
ContactUsLinkComponent
Triisopropylsilyl trifluoromethanesulphonate 97%
 
Triisopropylsilyl trifluoromethanesulphonate 97%
  ACRO250700100
 :  
undefined
 
Triisopropylsilyl trifluoromethanesulphonate 97%
  ACRO250700100
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    Clear colorless to yellow to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4130 to 1.4160 (20°C, 589 nm)
  • Titration with NaOH:
    ≥96.0 %

 

 

 :
 :306.42 g/mol
 :120 °C (1013 hPa)
 :1.14 g/cm³
 :100 °C
 :Ambient
 :3265
 :8,II
 :GHS05
 :80522-42-5
 :C₁₀H₂₁F₃O₃SSi
 :MFCD00009913