

1,1,1,3,3,3-Hexafluoro-2-propanol 99.5+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:920-66-1
:
Formula:C₃H₂F₆O
MDL Number:MFCD00011651
Merck Index:13,09598
Boiling Pt:59 °C (760 mmHg)
Melting Pt:–4 °C
Density:1.596 g/cm³
Flash Pt:>100 °C
UN:3265
ADR:8,I
:168.04 g/mol
EINECS:213-059-4
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.3 %
- GC:≥99.50 %
- Specific optical rotation:+170° to +180° (20°C, 589 nm) (C=2 in CHCl3)