Order Entry
China
ContactUsLinkComponent
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), crystalline powder USP, FCC, J.T.Baker®
Irritant
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), crystalline powder USP, FCC, J.T.Baker®
Catalog # BAKR0120-05
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), crystalline powder USP, FCC, J.T.Baker®
Catalog # BAKR0120-05
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Food GMP Manufactured Product:
    99.5 - 100.5%
  • GMP Manufactured Product:
    Passes Test
  • Meets F.C.C. Requirements:
    Passes Test
  • Meets U.S.P Requirements:
    Passes Test
  • Preserve in Tight Containers:
    7.5 - 9.0%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.05%
  • Bulk Food Chemical:
    ≤0.036%
  • Bulk Pharmaceutical Chemical:
    ≤0.015%
  • USP - Assay (C₆H₈O₇) (anhydrous basis):
    99.5 - 100.5 %
  • USP - Clarity of Solution:
    Passes Test
  • USP - Color of Solution:
    Passes Test
  • USP - Identification (USP):
    Passes Test
  • USP - Water (H₂O) (U.S.P.):
    7.5 - 9.0 %
  • USP - Residue on Ignition:
    ≤ 0.05 %
  • USP - Oxalic Acid (USP):
    ≤ 0.036 %
  • USP - Sulfate (SO₄):
    ≤ 0.015 %
  • USP - Readily Carbonizable Substances:
    Passes Test
  • FCC - Assay (C₆H₈O₇) (anhydrous basis):
    99.5 - 100.5 %
  • FCC - Identification (FCC):
    Passes Test
  • FCC - Lead (Pb):
    ≤ 0.5 mg/kg
  • FCC - Oxalate (C₂O₄) (FCC):
    ≤100 mg/kg
  • FCC - Readily Carbonizable Substances:
    Passes Test
  • FCC - Residue on Ignition:
    ≤ 0.05 %
  • FCC - Water (H₂O) (F.C.C.):
    ≤ 8.8 %

Specification Test Results

About this item

 :
 :210.14 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1.54 g/cm³
Flash Pt:173.9 °C (closed cup)
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
 :GHS07
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972