Glycine 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
:75.07 g/mol
Merck Index:13,04504
Boiling Pt:181 °C (1013 hPa)
Melting Pt:83 °C
Density:1.64 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-272-2
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification:Passes Test
- USP - Loss on Drying at 105°C:≤0.2 %
- USP - Residue on Ignition:≤0.1 %
- USP - Chloride (Cl):≤0.007 %
- USP - Sulfate (SO₄):≤0.0065 %
- USP - Hydrolyzable Substances:Passes Test
- USP - Assay (C₂H₅NO₂) (dried basis):98.5 - 101.5 %
- USP - Endotoxin Concentration (EU/g):≤2.5
- USP - Related Compounds: Glycine anhydride:≤0.1 %
- USP - Related Compounds: Monochloroacetic acid (For Information Only):Passes Test
- USP - Related Compounds: Iminodiacetic acid:≤0.1 %
- USP - Related Compounds: Diglycine:≤0.1 %
- USP - Related Compounds: Triglycine:≤0.1 %
- USP - Related Compounds: Hexamethylenetetramine:≤0.1 %
- USP - Related Compounds: Any unspecified impurity (For Information Only):= 0.10 %
- USP - Related Compounds: Total impurities:≤1.0 %
- EP/BP - Assay (C₂H₅NO₂) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH:5.9 - 6.4
- EP/BP - Chloride (Cl):≤75 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- EP/ BP - Related substances: Glycine anhydride:≤0.10 %
- EP/ BP - Related substances: Diglycine:≤0.10 %
- EP/ BP - Related substances: Triglycine:≤0.10 %
- EP/ BP - Related substances: Unspecified impurities, each:≤0.10 %
- EP/ BP - Related substances: Total impurities:≤0.2 %
- EP/BP - Ninhydrin Positive Substances - Each Impurity:≤0.10 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤1.0 %
- EP/BP - Ammonium (NH₄):≤0.02 %
- JP - Assay (C₂H₅NO₂) (dried basis):98.5 - 101.0 %
- JP - Identification:Passes Test
- JP - pH (1 in 20):5.6 - 6.6
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.021 %
- JP - Sulfate (SO₄):≤0.028 %
- JP - Ammonium (NH₄):≤0.02 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤0.30 %
- JP - Residue on Ignition:≤0.10 %
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKR0582
SDS
spec_BAKR0582-01 spec_BAKR0582-05 spec_BAKR0582-07 spec_BAKR0582-09