¥0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
China
Orders Link
ContactUsLinkComponent
Urea
Urea, Sigma-Aldrich®
Urea, Sigma-Aldrich®
Urea, Sigma-Aldrich®
Glass bottle for solids, 500 g
SIALU0631-500G
:
SIGMAALDRICH
Urea, Sigma-Aldrich®
SIALU0631-500G
:
SIGMAALDRICH
:
57-13-6
Expand image
Urea, Sigma-Aldrich®
Glass bottle for solids, 500 g
Add to cart
:
57-13-6
Unit of Measure
1 * 500 克
-
+
:
Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
:
57-13-6
:
NH₂CONH₂
:
MFCD00008022
:
60.06 g/mol
:
13,09935
:
197 °C (1013 hPa)
:
132.7 °C
:
1.34 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ200
:
Ambient
:
200-315-5
:
01-2119463277-33
Documents
Pack type:
Glass bottle for solids
Pk:
500 g