Order Entry
China
Orders LinkContactUsLinkComponent
Formamide ≥99.5%, Ultrapure

Formamide ≥99.5%, Ultrapure

Supplier:   VWR Chemicals
Formamide ≥99.5%, Ultrapure
Supplier:  VWR Chemicals
Certificates
Formamide ≥99.5%, Ultrapure

3 Options Available Below

Product Details & Documents
Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:13,04261
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:2 °C
  • Density:1.1334 g/cm³ (20 °C)
  • Flash Pt:79 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-842-0
Specifications for Product Family

Specification Test Results

  • Abs.@280 nm:
    ≤0.05
  • Conductivity:
    ≤100 umhos
  • Copper:
    <0.00001%
  • DNase:
    None Detected
  • Freezing Range:
    1 – 3 °C
  • Iron:
    <0.0005%
  • Lead:
    <0.00005%
  • Moisture (KF):
    <0.75%
  • Purity:
    >99.5%
  • RNase:
    None
  • Zinc:
    <0.00005%
Product Family Options

Product Information