Order Entry
China
ContactUsLinkComponent
 
N,N-Dimethylformamide, SupraSolv® for gas chromatography, Supelco®
Catalog # MERC1.10983.1000
Supplier:  MERCK MILLIPORE
CAS Number:  
undefined
N,N-Dimethylformamide, SupraSolv® for gas chromatography, Supelco®
Catalog # MERC1.10983.1000
Supplier:  MERCK MILLIPORE
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -61 °C
  • Molar Mass:
    73.09 g/mol
  • Boiling Point:
    153 °C (1013 hPa)
  • Vapor Pressure:
    3.77 hPa (20 °C)
  • Flash Point:
    67 °C
  • Refractive Index:
    1.43
  • Explosion Limit:
    2.2 - 16 %(V)
  • Density:
    0.94 g/cm3 (20 °C)
  • pH:
    7 (200 g/l, H2O, 20 °C)
  • Ignition Point:
    410 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

About this item

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
 :
 :73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,
 :M-Clarity™ quality level = MQ100
REACH:01-2119475605-32
 :GHS02
 :GHS08
 :GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284