Order Entry
China
ContactUsLinkComponent
Formaldehyde 37% (w/w) in aqueous solution stabilised
Toxic
Corrosive
Health
Formaldehyde 37% (w/w) in aqueous solution stabilised
Catalog # SIALF1635-500ML
Supplier:  SIGMAALDRICH
CAS Number:  
Toxic
Corrosive
Health
Formaldehyde 37% (w/w) in aqueous solution stabilised
Catalog # SIALF1635-500ML
Supplier:  SIGMAALDRICH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :30.03 g/mol
ADR:8,III
 :M-Clarity™ quality level = MQ100
 :Stabilised with methanol 10-15%
EINECS:200-001-8
UN:2209
 :GHS06
 :GHS05
 :GHS08
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274