2,2,2-Tribromoethanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-80-9
:
Formula:Br₃CCH₂OH
MDL Number:MFCD00004671
:282.76 g/mol
Merck Index:12,09743
Boiling Pt:92 - 93 °C (10 -rr)
Melting Pt:73…79 °C
Storage Temperature:Ambient
EINECS:200-903-1
:GHS07
Specification Test Results
- Appearance:White to yellow to light brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:73 to 82 °C
- GC:≥98.5 %