Order Entry
China
Orders LinkContactUsLinkComponent
Sorbic acid 99%

Sorbic acid 99%

Sorbic acid 99%
Certificates
Sorbic acid 99%

4 Options Available Below

Product Details & Documents
Synonyms:  (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
  • CAS Number:110-44-1
  • Formula:CH₃CH=CHCH=CHCOOH
  • MDL Number:MFCD00002703
  • Molecular Weight:112.13 g/mol
  • Merck Index:13,08793
  • Boiling Pt:228 °C (1013 hPa)
  • Melting Pt:132…135 °C
  • Density:1,204 g/cm³ (20 °C)
  • Flash Pt:127 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:203-768-7

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to cream Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    132 to 135 °C
  • Water:
    ≤0.5 % (K.F.)
  • Sulfated ash:
    ≤0.2 %
  • Titration with NaOH:
    98.5 to 101.5 % (On anhydrous substance)
  • Arsenic (As):
    ≤3 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
Product Family Options

Product Information