Supelco® Organic Reference Standard, Methyl linoleate
SUPL62280-5ML
: SUPELCO
:
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:112-63-0
- Description:Methyl linoleate
- Storage conditions:2…8 °C
- Hazard class description:H413
- Shipping conditions:STD
- MDL No.:MFCD00009534
- M-Clarity quality level:MQ100
- Synonym(s):Linoleic acid methyl ester; Methyl cis,cis-9,12-octadecadienoate
- Melting point:–35 °C (lit.)
- Formula:CH₃(CH₂)₃(CH₂CH=CH)₂(CH₂)₇CO₂CH₃
- Molecular weight:294,47
- Density:0,889 g/ml at 25 °C (lit.)
- Boiling point:192 °C/4 mmHg (lit.)
: P273 - P501