
(1S,2S)-(+)-1,2-Diaminocyclohexane ≥98.0% (by GC, titration analysis)
供应商: TCI
证书
关于此项目
CAS 编号:21436-03-3
:
化学式:C₆H₁₄N₂
MDL 编号:MFCD00062986
:114.19 g/mol
沸点:81°C
熔点:40…43 °C
储存温度:Refrigerator
UN:3259
ADR:8,III
:GHS05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
规格测试结果
- Appearance:White to Orange to Green powder to lump
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 98.0 ee%
- Purity (Nonaqueous Titration):min. 98.0 %