L(+)-Histidine USP, Multi-Compendial, J.T.Baker®
Catalog # BAKR2080-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:HDPE pail, Flowmor™
- Pk:12 kg
- Seq:4187
Specifications
Specification Test Results
- USP - Assay (C₆H₉N₃O₂) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation α²⁵D (+):12.6 - 14.0 Degree
- USP - pH:7.0 - 8.5
- USP - Loss on Drying at 105°C:≤ 0.2 %
- USP - Residue on Ignition:≤ 0.4 %
- USP - Chloride (Cl):≤ 0.05 %
- USP - Sulfate (SO₄):≤ 0.03 %
- USP - Iron (Fe):≤ 0.003 %
- USP - Related Compounds - Individual Impurities:≤ 0.5 %
- USP - Related Compounds - Total Impurities:≤ 2.0 %
- EP/BP - Assay (C₆H₉N₃O₂) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotationα²ºD (+):11.4 - 12.4 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤ 0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤ 0.5 %
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 300 ppm
- EP/BP - Ammonium (NH₄):≤ 0.02 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying:≤ 0.5 %
- EP/BP - Ash (sulfated):≤ 0.1 %
- Endotoxin Concentration, IU/mg ,For Information Only:= 200 ppm
- JP - Assay (C6H9N3O2) (dried basis):99.0 - 101.0 %
- JP - Ammonium (NH₄):≤ 0.02 %
- JP - Chloride (Cl):≤ 0.021 %
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Iron (Fe):≤ 10 ppm
- JP - Identification:Passes Test
- JP - Loss on Drying at 105°C:≤ 0.3 %
- JP- Optical Rotationα²ºD (+):11.8 - 12.8 Degree
- JP - Related Substances:Passes Test
- JP - pH:7.0 - 8.5
- JP - Residue on Ignition:≤ 0.10 %
- JP - Sulfate (SO₄):≤ 0.028 %
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:= 0.3 %
- Elemental Impurities (USP (232), EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.:11.8 - 12.8 Degree
Specification Test Results
About this item
:
:155.16 g/mol
Merck Index:13,04741
Melting Pt:272…273 °C
Density:1.44 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-745-3
CAS Number:71-00-1
Formula:C₆H₉N₃O₂
MDL Number:MFCD00064315