L-Arginine hydrochloride USP, Multi-Compendial, J.T.Baker®
Catalog # BAKR2067-07
Supplier: Avantor
CAS Number:
Specifications
- Pk:12 kg
- Seq:4187
Specifications
Specification Test Results
- USP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²º^D (+):21.4 - 23.6 Degree
- USP - Loss on Drying at 105°C:≤0.2 %
- USP - Residue on Ignition:≤0.1 %
- USP - Sulfate (SO₄):≤0.03 %
- USP - Related Compounds - Individual Impurities:≤0.5 %
- USP - Related Compounds - Total Impurities:≤2.0 %
- USP - Chloride (Cl):16.5 - 17.1 %
- EP/BP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):21.0 - 23.5 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤0.5 %
- EP/BP - Sulfate (SO₄):≤300 ppm
- EP/BP - Ammonium (NH₄):≤0.02 %
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis):98.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Optical Rotation (+):21.5 - 23.5 Degree
- JP - pH (1 in 10):4.7 - 6.2
- JP - Clarity and Color of Solution:Passes Test
- JP - Sulfate (SO₄):≤0.028 %
- JP - Ammonium (NH₄):≤0.02 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤0.20 %
- JP - Residue after Ignition:≤0.10 %
- Endotoxin Concentration, IU/mg, For Information Only:≤ 0.20 %
- No Class 1,2,3 or other solvents are used or produced in the:≤ 0.10 %
- manufacturing or purification of the product.:= 2 ppm
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:Passes Test
- Elemental Impurities (USP (232), EP 5.20) - Information on elemental:= 0.20 %
- impurities for this product is available on the associated Product:= 0.10 %
- Regulatory Data Sheet and elemental impurity profile report.:For Information Only
Specification Test Results
About this item
:
:210.66 g/mol
Merck Index:13,00785
Melting Pt:218 °C
Density:1.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-275-1
CAS Number:1119-34-2
Formula:C₆H₁₅ClN₄O₂
MDL Number:MFCD00064550