Order Entry
China
ContactUsLinkComponent
Nickel(II) nitrate hexahydrate 99% for analysis
Nickel(II) nitrate hexahydrate 99% for analysis
Catalog # ACRO223155000
CAS Number:  
undefined
Nickel(II) nitrate hexahydrate 99% for analysis
Catalog # ACRO223155000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Green to blue-green Crystals and/or chunks
  • Titration Complexometric:
    ≥98.5 %
  • Chloride (Cl):
    ≤50 ppm
  • Aluminium (Al):
    ≤10 ppm
  • Arsenic (As):
    ≤5 ppm
  • Cadmium (Cd):
    ≤5 ppm
  • Calcium (Ca):
    ≤20 ppm
  • Chromium (Cr):
    ≤5 ppm
  • Cobalt (Co):
    ≤500 ppm
  • Copper (Cu):
    ≤10 ppm
  • Iron (Fe):
    ≤10 ppm
  • Lead (Pb):
    ≤5 ppm
  • Magnesium (Mg):
    ≤20 ppm
  • Manganese (Mn):
    ≤20 ppm
  • Sulfur (S):
    ≤100 ppm
  • Zinc (Zn):
    ≤10 ppm
  • Insoluble matter:
    ≤0.01 %

Specification Test Results

About this item

 :
 :290.79 g/mol
Merck Index:12,06596
Boiling Pt:137 °C (760 mmHg)
Melting Pt:56.7 °C
Density:2,05 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-068-5
UN:2725
ADR:5.1,III
 :GHS03
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:13478-00-7
Formula:Ni(NO₃)₂·6H₂O
MDL Number:MFCD00149805