Order Entry
China
ContactUsLinkComponent
Cobalt(II) nitrate hexahydrate ≥98% ACS, Fluka™
Oxidizing
Health
Corrosive
Irritant
Cobalt(II) nitrate hexahydrate ≥98% ACS, Fluka™
Supplier:  HONEYWELL
Certificates
Oxidizing
Health
Corrosive
Irritant
Cobalt(II) nitrate hexahydrate ≥98% ACS, Fluka™
Supplier:  HONEYWELL

Specification Test Results

  • Appearance of the substance:
    complying
  • Assay:
    98.0 - 102.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 0.005 %
  • Copper (Cu):
    Max. 0.002 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.01 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.05 %
  • Nickel (Ni):
    Max. 0.15 %
  • Lead (Pb):
    Max. 0.002 %
  • Zinc (Zn):
    Max. 0.01 %
  • Chloride (Cl):
    Max. 0.002 %
  • Sulfate (SO₄):
    Max. 0.005 %

Specification Test Results

Related Information