Order Entry
China
ContactUsLinkComponent
Cobalt(II) chloride hexahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Health
Irritant
Environment
Cobalt(II) chloride hexahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Catalog # MERC1.02539.0100
Supplier:  MERCK MILLIPORE
CAS Number:  
Health
Irritant
Environment
Cobalt(II) chloride hexahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Catalog # MERC1.02539.0100
Supplier:  MERCK MILLIPORE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    191 g/l (100 °C)
  • Melting Point:
    56 °C
  • Molar Mass:
    237.90 g/mol
  • Bulk Density:
    1250 kg/m3
  • Density:
    1.92 g/cm3 (25 °C)
  • pH:
    4.9 (50 g/l, H2O, 25 °C)
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material

Specification Test Results

About this item

 :
 :237.93 g/mol
Merck Index:13,02462
Boiling Pt:110 °C (1013 hPa)
Melting Pt:56 °C
Density:1.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-589-4
UN:3077
ADR:9,III
 :M-Clarity™ quality level = MQ300
 :GHS08
 :GHS07
 :GHS09
CAS Number:7791-13-1
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652