Order Entry
China
ContactUsLinkComponent
Chromium(III) chloride hexahydrate 98%
Irritant
Corrosive
Environment
Chromium(III) chloride hexahydrate 98%
Catalog # ACRO213365000
CAS Number:  
undefined
Irritant
Corrosive
Environment
Chromium(III) chloride hexahydrate 98%
Catalog # ACRO213365000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    Dark green Crystalline powder
  • Titration with Na2S2O3:
    ≥97.5 %
  • Sulfate (SO4):
    ≤500 ppm
  • Iron (Fe):
    ≤300 ppm

Specification Test Results

About this item

 :
 :266.45 g/mol
Merck Index:13,02242
Melting Pt:83 °C
Density:1,76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-038-3
UN:3260
ADR:8,III
 :GHS07
 :GHS05
 :GHS09
CAS Number:10060-12-5
Formula:CrCl₃·6H₂O
MDL Number:MFCD00149660