


Zinc sulphate heptahydrate Ph. Eur., USP
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7446-20-0
:
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00149894
Merck Index:12,10293
Melting Pt:100 °C
Density:1.97 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-793-3
UN:3077
ADR:9,III
:287.54 g/mol
:GHS07
:GHS05
:GHS09
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Conforms (see CoA/CoS)
- Residual solvents:Unlikely by manuf.process
- Cd (Cadmium):Max. 1 ppm
- Co (Cobalt):Max. 1 ppm
- Hg (Mercury):Max. 3 ppm
- Ni (Nickel):Max. 3 ppm
- V (Vanadium):Max. 1 ppm
- Conforms to Ph.Eur./USP:Passes test
- Ph.Eur. specifications::Max. 100 ppm
- Assay [Ph.Eur.]:99.0 --> 104.0 %
- Identification A:Passes test Ph.Eur.
- Identification B:Passes test Ph.Eur.
- Identification C:Passes test Ph.Eur.
- Appearance of solution:Passes test Ph.Eur.
- pH (5 %) [Ph.Eur.]:4.4 --> 5.6
- Cl (Chloride) [Ph.Eur.]:Max. 300 ppm
- Fe (Iron) [Ph.Eur.]:Max. 100 ppm
- Assay [USP]:99.0 --> 108.7 %
- As (Arsenic) [USP]:Max. 14 ppm
- Pb (Lead) [USP]:Max. 20 ppm
- Alkalies and alkaline earths [USP]:Max. 0.9 %
- Acidity:Passes test USP
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
Packaging Information
BSE_29158_ZINC SULPHATE 7H2O PH.EUR
BSE/TSE Certificates
spec_VWRC29158.294 spec_VWRC29158.363 spec_VWRC29158.363
Product specification
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Zinc Sulphate Heptahydrate Ph. Eur., USP