


Copper(II) nitrate trihydrate ≥98%, GPR RECTAPUR®
Catalog # VWRC23123.294
供应商: VWR Chemicals
CAS 编号:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
规格
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
规格
规格测试结果
- Assay:Min. 98 %
- Cl (Chloride):Max. 0.02 %
- SO4 (Sulphate):Max. 0.05 %
- Fe (Iron):Max. 100 ppm
- Pb (Lead):Max. 100 ppm
规格测试结果
关于此项目
:
:241.6 g/mol
熔点:114 °C
密度:2.05 g/cm³ (20 °C)
EINECS:221-838-5
UN:1477
ADR:5.1,II
:GHS03
:GHS07
:GHS09
CAS 编号:10031-43-3
化学式:Cu(NO₃)₂·3H₂O
MDL 编号:MFCD00149671