Magnesium sulphate heptahydrate 99.0-100.5%, GenAR® USP for parenteral use, Macron Fine Chemicals™
Catalog # MACR5691-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Plastic bag
- Pk:100 lb
- Seq:7167
- Environmentally Preferable:
Specifications
Specification Test Results
- GMP Manufactured Product:—
- Meets B.P. Chemical Specifications:—
- Meets E.P. Chemical Specifications:—
- Meets U.S.P Requirements:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:—
- USP - Assay (as MgSO₄):99.0 - 100.5 %
- Appearance:Passes Test
- Solubility (5 in 50):Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - pH:5.0 - 9.2
- USP - Loss on Ignition:40.0 - 52.0 %
- USP - Chloride (Cl):≤ 0.014 %
- USP - Selenium (Se):≤ 0.003 %
- USP - Iron (Fe):≤ 0.5 ppm
- Assay (MgSO₄):99.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Chloride (Cl):≤ 300 ppm
- EP - Iron (Fe):≤ 20 ppm
- EP/BP - Loss on Drying:48.0 - 52.0 %
- Elemental Impurities (USP (232), EP 5.20) - Information on:48.0 - 52.0 %
Specification Test Results
About this item
Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
:
:246.48 g/mol
Melting Pt:1124
Density:2.66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:10034-99-8
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785
Merck Index:13,05715
:Management of Change (MOC) category = R