Magnesium sulphate heptahydrate 99+%, extra pure
: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
:10034-99-8
:
:MgSO₄·7H₂O
:MFCD00149785
:246.48 g/mol
:13,05715
:770 °C
:2,66 g/cm³ (20 °C)
:Ambient
:231-298-2
- Environmentally Preferable:
- Appearance:White Crystalline powder or crystals
- Titration Complexometric:≥99.0 % (on dry substance)
- Loss on drying:48.0 % to 52.0 % (450°C)
- pH:5.0 to 9.2 (5 % at 20°C)
- Chloride (Cl):≤200 ppm
- Heavy metals (as Pb):≤10 ppm