Aluminium potassium sulphate dodecahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
目录编号 MERC1.01047.1000
Supplier: MERCK MILLIPORE
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Solubility:139 g/l (20 °C)
- Melting Point:92.5 °C
- Molar Mass:474.39 g/mol
- Bulk Density:900 kg/m3
- Density:1.75 g/cm3 (20 °C)
- pH:3.0 - 3.5 (100 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:474.39 g/mol
Merck Index:12,00373
Melting Pt:92.5 °C
Density:1.72 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:233-141-3
CAS Number:7784-24-9
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143