
Potassium hexacyanoferrate(II) trihydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Catalog # MERC1.04984.0100
Supplier: MERCK MILLIPORE
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:289 g/l (20 °C)
- Melting Point:70 °C Elimination of water of crystallisation
- Molar Mass:422.39 g/mol
- Bulk Density:950 - 1050 kg/m3
- Density:1.85 g/cm3 (20 °C) (anhydrous substance)
- pH:9.5 (100 g/l, H2O, 20 °C) (anhydrous substance)
Specification Test Results
About this item
:
:422.39 g/mol
Merck Index:12,07793
Melting Pt:70 °C
Density:1.889 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:237-722-2
:GHS09
Storage Temperature:Ambient
CAS Number:14459-95-1
Formula:K₄Fe(CN)₆·3H₂O
MDL Number:MFCD00167023