

di-Sodium tetraborate decahydrate 99.0-105.0% NF, J.T.Baker®
供应商: Avantor
证书
关于此项目
CAS 编号:1303-96-4
:
化学式:Na₂B₄O₇·10H₂O
MDL 编号:MFCD00149193
:381.37 g/mol
默克索引:13,08662
沸点:320 °C (1013 hPa)
熔点:75 °C
密度:1.73 g/cm³
储存温度:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS08
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
规格测试结果
- Meets N.F. Requirements:99.0 - 105.0%
- GMP Manufactured Product:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:Passes Test
- NF - Assay:99.0 - 105.0 %
- NF - Identification:Passes Test
- NF - Carbonate and Bicarbonate:Passes Test