tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystals USP, FCC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:6132-04-3
:
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031
:294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Food GMP Manufactured Product:99.0 - 100.5%
- Meets F.C.C. Requirements:Passes Test
- Meets U.S.P Requirements:Passes Test
- GMP Manufactured Product:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:10.0 - 13.0%
- Bulk Food Chemical:Passes Test
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Assay (anhydrous basis):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - Water (H₂O):10.0 - 13.0 %
- USP - Alkalinity:Passes Test
- USP - Tartrate:Passes Test
- FCC - Assay (anhydrous basis):99.0 - 100.5 %
- FCC - Identification:Passes Test
- FCC - Alkalinity:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Water (H₂O):10.0 - 13.0 %
- Solubility (5 in 50):Passes Test