Order Entry
China
ContactUsLinkComponent
Magnesium acetate tetrahydrate analytical reagent
Magnesium acetate tetrahydrate analytical reagent
Catalog # VWRC0131-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Magnesium acetate tetrahydrate analytical reagent
Catalog # VWRC0131-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Chloride:
    ≤0.01%
  • Insolubles:
    ≤0.005%
  • Iron:
    ≤0.01%
  • Sulphate:
    ≤0.05%

Specification Test Results

About this item

 :
 :214.46 g/mol
Merck Index:13,05676
Melting Pt:80 °C
Density:1.454 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-554-9
CAS Number:16674-78-5
Formula:(H₃CCOO)₂Mg·4H₂O
MDL Number:MFCD00149214