Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), 4 L
Catalog # INVT15558026
Marketsource
Supplier: INVITROGEN
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Catalog # INVT15558026
Supplier: INVITROGEN
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:4 L
- Description:TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer)



