

Ammonia solution 20%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Density:0.923 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay:18.0 - 22.0 %
- Colouration:Max. 10 APHA
- Density (20/4):0.910 - 0.930
- Evaporation residue:Max. 10 ppm
- Silica:Max. 10 ppm
- Substances reducing KMnO4 (as O):Max. 4 ppm
- Cl (Chloride):Max. 0.5 ppm
- CO3 (Carbonate):Max. 10 ppm
- PO4 (Phosphate):Max. 0.5 ppm
- S (Sulphur):Max. 0.2 ppm
- SO4 (Sulphate):Max. 20 ppm
- Al (Aluminium):Max. 0.03 ppm
- As (Arsenic):Max. 0.01 ppm
- B (Boron):Max. 0.05 ppm
- Ba (Barium):Max. 0.02 ppm
- Be (Beryllium):Max. 0.02 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.5 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.3 ppm
- Li (Lithium):Max. 0.01 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.01 ppm
- Mo (Molybdenum):Max. 0.02 ppm
- Na (Sodium):Max. 0.8 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.02 ppm
- Sr (Strontium):Max. 0.01 ppm
- Ti (Titanium):Max. 0.1 ppm
- V (Vanadium):Max. 0.01 ppm
- Zn (Zinc):Max. 1 ppm
- Zr (Zirconium):Max. 0.1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
spec_VWRC21188.294 spec_VWRC21188.294 spec_VWRC21188.363