


Tetramethylammonium hydroxide 10% in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-59-2
:
Formula:C₄H₁₃NO
MDL Number:MFCD00008280
:91.15 g/mol
Boiling Pt:110 °C (1013 hPa)
Density:1 g/cm³
EINECS:200-882-9
UN:1835
ADR:8,II
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Solution
- Titration with HCl:9.5 to 10.5 %
- Refractive index:1.6020 to 1.6050 (20°C, 589 nm)
- GC:≥97.5 %